missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Solvent CM, Reagent for volumetric two-component KF titration in oils (working medium), Honeywell Fluka™
104.92 € - 745.25 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15694460
|
Honeywell-Hydranal
34812-1L |
1 L |
104.92 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15604470
|
Honeywell-Hydranal
34812-2.5L |
2.5 L |
220.03 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624470
|
Honeywell Fluka
34812-6X1L |
6 x 1 L |
531.34 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15614470
|
Honeywell Fluka
34812-4X2.5L |
4 x 2.5 L |
745.25 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
MFCD00147198 | |
UN2810 |
Safety and Handling
P201-P260-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Solvent CM
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H331-Toxic if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trichloromethane; chloroform, methanol,imidazole, sulphur dioxide