missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Solvent E, Honeywell Fluka™

Medium for volumetric two-component Karl Fischer titration (ethanol-based)

60.41 € - 711.11 €

Specifications

Density 0.90g/mL
Flash Point 12°C (53.6°F)
Chemical Name or Material HYDRANAL™-Solvent E
CAS 64-17-5, 111-42-2, 288-32-4, 7446-09-5
MDL Number MFCD01867183
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15624370
View Documents
Honeywell-Hydranal
34730-500ML
500 mL
60.41 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15694360
View Documents
Honeywell-Hydranal
34730-1L
1 L
99.51 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15604370
View Documents
Honeywell-Hydranal
34730-2.5L
2.5 L
204.55 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15644370
View Documents
Honeywell-Hydranal
34730-6X500ML
6 x 500 mL
315.46 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15634370
View Documents
Honeywell-Hydranal
34730-6X1L
6 x 1 L
519.28 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15614370
View Documents
Honeywell-Hydranal
34730-4X2.5L
4 x 2.5 L
711.11 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

Specifications

Specifications

0.90g/mL
HYDRANAL™-Solvent E
MFCD01867183
12°C (53.6°F)
64-17-5, 111-42-2, 288-32-4, 7446-09-5
UN1170
Videos
Safety and Handling

Safety and Handling

P201-P210-P280-P308 + P313-P370 + P378-P403 + P235

Product Identifier
  • HYDRANAL Solvent E

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, 2,2'-iminodiethanol; diethanolamine,imidazole, sulphurdioxide

SDS
Documents

Documents

Product Certifications