missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5, Honeywell Fluka™

Product Code. p-7203375 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
6 L
This item is not returnable. View return policy

Product Code. 16821910

Brand: Honeywell Fluka 348011LOM

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titrating agent for volumetric, two-component Karl Fischer titrations (Methanol-based)

Advantages of Hydranal™ Two-Component Reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents offer reduced toxicity compared to methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Titrant 5
Melting Point -98°C
Boiling Point 65°C (at 1.013 hPa)
Color Brown
Density 0.850 g/cm3 (at 20°C)
pH Neutral
Physical Form Liquid
CAS 7553-56-2
Quantity 1 L
Flash Point 11°C
Packaging Metrohm OMNIS Smart Reagent
UN Number UN1230
Synonym Methyl Alcohol
Solubility Information Completely Miscible in Water
Grade Volumetric Two-Component KF Titration
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL NEXTGEN Titrant 5 OM
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, iodine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.