Learn More
HYDRANAL™ - Titrant 5 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™
Titrating agent for volumetric two-component Karl Fischer titration (ethanol-based)
52.47 € - 340.45 €
Specifications
UN Number UN1170
View More Specs
Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15654370
View Documents
Honeywell-Hydranal
34732-100ML
100 mL
52.47 €
100mL
15694370
View Documents
Honeywell-Hydranal
34732-500ML
500 mL
62.35 €
500mL
15664370
View Documents
Honeywell-Hydranal
34732-1L
1 L
103.73 €
1 L
15674370
View Documents
Honeywell-Hydranal
34732-2.5L
2.5 L
214.33 €
2.50 L
15614380
View Documents
Honeywell-Hydranal
34732-6X500ML
6 x 500 mL
340.45 €
Pack of 6
Description
Description
Advantages of Hydranal two-component reagents:
  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)
Improved Composition
In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.
Specifications
Specifications
UN1170
Videos
Safety and Handling
Safety and Handling
P210-P260-P305 + P351 + P338-P370 + P378-P403 + P235
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
HYDRANAL™ - Titrant 5 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™