missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Water Standard KF-Oven, 150-160°C, Honeywell Fluka™

Standard for Karl Fischer oven check (water content ∼5.0%, exact value on report of analysis)

Brand:  Honeywell-Hydranal 34693-10G

45.49 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.



Product Code. 15624350

  • 47.88 € / 10g

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Advantages of HYDRANAL-Water Standards:

  • Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
  • Manufactured according to ISO 9001 requirements
  • Report of Analysis showing the exact water content and expanded uncertainty

Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222

  • Long shelf life (up to five years)
  • Convenient packaging
  • Supplied with detailed instruction for use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYDRANAL™-Water Standard KF-Oven, 150-160°C
10 g
64044-51-5
NONH for all modes of transport
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : trichloromethane; chloroform, 1-methoxy-2-propanol; monopropylene glycol methylether, 2,2,2-Trifluo

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.