Learn More
3M™ Hydrated-Sponge
Uses simplified packaging to minimize handling and preparation, and reduces waste.
Brand:  3M™ 7100067833
Encompass
 View more versions of this product
Product Code. 17203774
  • POA / 100 sets
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection.
  • Durable sponge withstands scrubbing.
  • Sponge bag with write-on label for ease in labeling samples.
  • Includes two decontaminated gloves for safety and convenience.
Specifications
Specifications
Hydrated Sponge
7.6 cm
4 cm
10 mL Buffered Peptone Water Broth and Gloves HS10BPW2G
3 in.
1.5 in.
100/Cs.