Learn More
Juchheim Laborgeraete™ Hydraulic Laboratory Jack
Use these hydraulic lifting stages for easy, safe lifting. Juchheim Laborgeraete™ Hydraulic Laboratory Jacks feature robust, light aluminum alloy construction.
Brand:  Juchheim Laborgeraete™ 02297/A
Product Code. 10444501
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Complete with pedal and anti-sag device   
  • Aluminum alloy construction
  • Stable, parallel movement
Specifications
Specifications
Hydraulic Laboratory Jack
Easy, safe lifting
230 mm
200 mm
230 x 200 x 91 mm
91 mm (min.), 285 mm (max.)
25 kg