missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals

47.13 € - 506.52 €

Chemical Identifiers

CAS 5341-61-7
Molecular Formula ClH5N2
Molecular Weight (g/mol) 68.50
MDL Number MFCD00064543
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
PubChem CID 17548
IUPAC Name hydrazine;dihydrochloride
SMILES [H+].[Cl-].NN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10371301
View Documents
Thermo Scientific Acros
411761000
100 g Glass bottle
47.13 €
100g
Please to purchase this item. Need a web account? Register with us today!
10762592
View Documents
Thermo Scientific Acros
411765000
500 g Glass bottle
148.74 €
500g
Please to purchase this item. Need a web account? Register with us today!
10697564
View Documents
Thermo Scientific Acros
411760025
2.5 kg Plastic Bottle
506.52 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

5341-61-7
68.50
BIVUUOPIAYRCAP-UHFFFAOYSA-N
17548
[H+].[Cl-].NN
ClH5N2
MFCD00064543
hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
hydrazine;dihydrochloride

Specifications

198.0°C
White
98.5% min. (Argentometry)
NH2NH2·2HCl
15, 4809
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
[H+].[Cl-].NN
68.50
104.97
Authentic
5341-61-7
Chunks and/or Powder
ClH5N2
MFCD00064543
hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
BIVUUOPIAYRCAP-UHFFFAOYSA-N
hydrazine;dihydrochloride
17548
99%
Hydrazine dihydrochloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Gently

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 226-283-2

missing translation for 'rtecsNumber' : MV2298000

missing translation for 'tsca' : TSCA

Product Identifier
  • Hydrazine dihydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Skin sensitiser Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Hydrazine, dihydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only