missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine hydrate, 100% (Hydrazine, 64%), Thermo Scientific Chemicals

26.03 € - 57.69 €

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine;hydrate
SMILES NN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10246540
View Documents
Thermo Scientific Acros
196711000
100 g Glass bottle
26.03 €
100g
Please to purchase this item. Need a web account? Register with us today!
10276440
View Documents
Thermo Scientific Acros
196715000
500 g Glass bottle
57.69 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

10217-52-4
32.05
OAKJQQAXSVQMHS-UHFFFAOYSA-N
24654
hydrazine;hydrate
H4N2
MFCD00149931
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
CHEBI:35511
NN

Specifications

120.1°C
10217-52-4
Liquid
H2NNH2·XH2O
01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174
14, 4771
Solubility in water: miscible. Other solubilities: miscible with alcohol, insoluble in chloroform and ether
NN
32.05
CHEBI:35511
100.00 to 100.93% (N2H4.xH2O), 64.00 to 64.60% (N2H4)
1.0315
Hydrazine hydrate, Hydrazine Hydrate 100% (Hydrazine, 64%)
-51.5°C
Colorless
H4N2
MFCD00149931
75°C
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
OAKJQQAXSVQMHS-UHFFFAOYSA-N
hydrazine;hydrate
24654
32.04
1.4285 to 1.4315
1.0315g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause cancer.
Very toxic to aquatic life with l

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre

GHS Signal Word: Danger

missing translation for 'rtecsNumber' : MV8050000

Product Identifier
  • Hydrazine hydrate, 100% (Hydrazine, 64%)

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Hydrazine (hydrate),Hydrazine

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.