missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Hydrate, 99+%, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Description
Hydrazine Hydrate, 99+%, Certified AR for Analysis
Specifications
Specifications
Boiling Point | 118°C |
Melting Point | -51.7°C/-61.1°F |
Color | Colorless |
pH | 12 |
Physical Form | Liquid |
Quantity | 100 mL |
Formula Weight | 50.06g/mol |
Packaging | Plastic Coated Glass bottle |
Safety and Handling
Product Identifier
- Hydrazine hydrate
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H350-May cause cancer.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction