missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine monohydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11377198
Click to view available options
Quantity:
100 g
500 g
2 kg
4 x 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Hydrazine Hydrate used as a reactant in the cyclizations of pyridinones. It is also used in the study of nanocrystal semiconductors, participating in the functionalization and passivation of surface states. It is widely used as a reducing agent or an intermediate of synthesis in various industrial sectors like water treatment (effluents, industrial boilers), chemical treatment process (metals, mine extraction) or active ingredients synthesis (pharmaceuticals and agrochemicals).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-57-8
Molecular Formula H6N2O
Molecular Weight (g/mol) 50.06
MDL Number MFCD00149931
InChI Key IKDUDTNKRLTJSI-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
SMILES O.NN

Specifications

Boiling Point 120°C to 121°C
Melting Point -52°C
Color Colorless
Physical Form Liquid
Quantity 2 kg
Assay Percent Range >99%
Linear Formula H2NNH2·H2O
UN Number 2030
Flash Point 75° (167°F)
Merck Index 14,4771
Solubility Information Soluble in chloroform,DMSO,methanol. Slightly soluble in water.
IUPAC Name hydrazine;hydrate
Formula Weight 50.06 (32.05 Anhydrous)
Percent Purity ≥99%
Odor Amine-like
Refractive Index 1.4310
Density 1.027g/mL
Chemical Name or Material Hydrazine Monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'