missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrindantin dihydrate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >240°C |
Color | Beige to Beige-Pink |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min |
Packaging | Glass bottle |
Quantity | 10 g |
Merck Index | 14, 4775 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 358.29 |
Percent Purity | 96% |
Show More |
Safety and Handling

Product Identifier
- Hydrindantin dihydrate
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction