missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic Acid, 55 to 60%, Honeywell™

Product Code. 15675710
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 10034-85-2
Density 1.7g/mL at 20°C
Packaging Glass bottle
Quantity 250 mL
Molecular Formula HI
MDL Number MFCD00011347
UN Number UN1787
Synonym hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILES I
IUPAC Name hydrogen iodide
Molecular Weight (g/mol) 127.91
PubChem CID 24841
ChEBI CHEBI:43451
Formula Weight 127.91g/mol
Percent Purity 55 to 60%
Chemical Name or Material Hydriodic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.