missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic acid, 57% w/w aqueous solution stabilized with 1.5% hypophosphorus acid, Thermo Scientific Chemicals

Product Code. 11398415
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Hydriodic acid is used to convert primary alcohol into alkyl iodide. It is used in organic and inorganic synthesis as one of the primary sources of iodine as well as a reducing agent. It is also used to cleave the ethers to get alkyl iodides and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10034-85-2, 7732-18-5, 6303-21-5
Melting Point -50.8°C
Boiling Point 126°C
CAS Max % 57, 41.5, 1.5
Density 1.701 g/mL
Packaging Plastic
Quantity 500 g
Molecular Formula HI
MDL Number MFCD00011347
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILES I
IUPAC Name hydrogen iodide
Molecular Weight (g/mol) 127.91
Chemical Name or Material Hydriodic acid
Show More Show Less
compliance-icons
Product Identifier
  • Hydriodic acid, 57%, stabilized with 1.5% hypophosphorous acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hydriodic acid, Hypophosphorus acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.