missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic acid, ACS reagent, 55% in water, without stabilizer, Thermo Scientific Chemicals

Product Code. 13247059
Click to view available options
Quantity:
50 mL
250 mL

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-85-2
Molecular Formula HI
Molecular Weight (g/mol) 127.91

Specifications

Boiling Point 127.0°C
Density 1.7000 g/mL
Packaging Glass bottle
Specific Gravity 1.7
Quantity 50 mL
Linear Formula HI
Fieser 01,449; 02,213; 06,281; 07,173; 08,246; 09,238; 10,199; 11,255; 15,166
Merck Index 15, 4814
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 127.91
Grade ACS Reagent
Chemical Name or Material Hydriodic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.