missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 11492644
Click to view available options
Quantity:
100 mL
Packaging:
Amber glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

pH 1
Color Brown-Red
CAS 10034-85-2
Melting Point -51°C
Boiling Point 127°C
Packaging Amber glass bottle
Quantity 100 mL
Molecular Formula HI
Physical Form Liquid
MDL Number MFCD00011347
Synonym hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group
InChI Key XMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILES I
IUPAC Name hydrogen iodide
Molecular Weight (g/mol) 127.91
PubChem CID 24841
ChEBI CHEBI:43451
Show More Show Less
compliance-icons
Product Identifier
  • Hydriodic acid SLR
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydriodic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.