missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles

Provides foot with side support and extra support at heel and middle of foot.

Brand:  Emma Safety Footwear MM00107241

 View more versions of this product

Product Code. 10372410

  • 34.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Regulates moisture
  • Contains active carbon to prevent odor
Specifications

Specifications

Gray
41
Stability Inlay Sole
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers