missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Hydro-Tec™ Stability Inlay Soles

Product Code. 10705092
Click to view available options
Size:
39
41
43
44
45
46
Unit Size:
1 pair
Each
This item is not returnable. View return policy

Product Code. 10705092

Brand: Emma Safety Footwear MM00107246

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides foot with side support and extra support at heel and middle of foot.

  • Regulates moisture
  • Contains active carbon to prevent odor

Specifications

Color Gray
Size 46
Product Type Stability Inlay Sole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.