missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, 48%, Certified AR for Analysis, Fisher Chemical™

Product Code. 10550531
Click to view available options
Quantity:
500 mL
2.5 L

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

pH lt;1
Color Yellow
Melting Point -11°C
Boiling Point 126°C
Packaging HDPE plastic bottle
Vapor Pressure 8mm Hg 25
Quantity 500 mL
Physical Form Liquid
IUPAC Name bromane
Formula Weight 80.9g/mol
compliance-icons compliance-icons
Product Identifier
  • Hydrobromic acid, ca. 48 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.