missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid, ≥99.99%; Honeywell Fluka™

Product Code. 15664100 Shop All Honeywell Fluka Products
500 mL, Glass Bottle
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

48 wt. % in H2O,≥99.99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

pH Acidic
Color Yellow
Composition 48 wt.% in water
Melting Point -11°C
Boiling Point 126°C
Density 1.49g/mL at 25°C
Odor Stinging
Packaging Glass Bottle
Vapor Pressure 320psi (21.1°C), 8mmHg (25°C)
Quantity 500 mL
UN Number UN1788
Beilstein 3587158
Solubility Information Completely miscible in water
IUPAC Name bromane
Formula Weight 80.91g/mol
Percent Purity ≥99.99%
Chemical Name or Material Hydrobromic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrobromic Acid, ≥99.99%; Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.