missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11351588
Click to view available options
Quantity:
5 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Air Sensitive

Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6, 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.49
Sensitivity Air Sensitive
Vapor Pressure 22500 hPa (16876mm Hg)
Assay Percent Range 99.9999% (metals basis)
Quantity 5 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible with water.
IUPAC Name hydrogen bromide
Formula Weight 80.92
Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.