missing translation for 'onlineSavingsMsg'
Learn More
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals Product Code.: 11374217

Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals

Product Code. 11374217
250 mL
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11374217

Brand: Thermo Scientific Alfa Aesar 036694.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Air Sensitive

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes, and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration, and dehydration, and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6, 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.49 g/cm3
Sensitivity Air Sensitive
Vapor Pressure 22500 hPa (16876mm Hg) at 20°C
Assay Percent Range 47.0-49.0%
Quantity 250 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible in water.
IUPAC Name hydrogen bromide
Formula Weight 80.92
Grade ACS
Concentration or Composition (by Analyte or Components) Hydrogen bromide: 48.0%; Water: 52.0%
Chemical Name or Material Hydrobromic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrobromic acid, ACS, 47.0-49.0%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrobromic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrobromic acid, ACS, 47.0-49.0%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.