missing translation for 'onlineSavingsMsg'
Learn More
Hydrobromic Acid, ACS Reagent, Honeywell Fluka™ Product Code.: 15682650

Hydrobromic Acid, ACS Reagent, Honeywell Fluka™

Product Code. 15682650 Shop All Honeywell Fluka Products
1 L
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 15682650

Brand: Honeywell-Fluka 2442601L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent, 48%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

Concentration 48%
Density 1.49g/mL at 25°C
Residue after Ignition ≤0.002%
Vapor Pressure 8mmHg (25°C), 320psi (21.1°C)
Quantity 1 L
UN Number 1788
Beilstein 3587158
IUPAC Name bromane
Formula Weight 80.91g/mol
Grade ACS Reagent
Chemical Name or Material Hydrobromic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrobromic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydrobromic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrobromic Acid, ACS Reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.