missing translation for 'onlineSavingsMsg'
Learn More
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals Product Code.: 11964101

Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals

Product Code. 11964101
2.5 L, Glass bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11964101

Brand: Thermo Scientific Acros 223320025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6, 7732-18-5
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name hydrogen bromide
SMILES Br

Specifications

Color Yellow
Melting Point -11.0°C
Boiling Point 126.0°C to 128.0°C
Density 1.4800g/mL
Heavy Metals (as Pb) 5ppm max.
Packaging Glass bottle
Specific Gravity 1.48
Quantity 2.5 L
Linear Formula HBr
Physical Form Solution
Fieser 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
Solubility Information Solubility in water: soluble.
Formula Weight 80.9
Percent Purity 47 to 49% (Argentometry)
Grade Analytical
Chemical Name or Material Hydrobromic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrobromic acid, ca. 48 wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.