missing translation for 'onlineSavingsMsg'
Learn More
Hydrobromic Acid, purum p.a., Honeywell™ Product Code.: 15651690

Hydrobromic Acid, purum p.a., Honeywell™

Product Code. 15651690 Shop All Honeywell Research Chemicals Products
1 L
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy

Product Code. 15651690

Brand: Honeywell Research Chemicals 187301L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purum p.a.,≥48%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromideShow MoreShow Less
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br

Specifications

Density 1.49g/mL at 25°C
Vapor Pressure 8mmHg (25°C), 320psi (21.1°C)
Quantity 1 L
UN Number 1788
Beilstein 3587158
Formula Weight 80.91g/mol
Percent Purity ≥48%
Grade purum p.a.
Chemical Name or Material Hydrobromic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrobromic Acid, purum p.a., Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.