Learn More
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266
76.80 € - 258.00 €
Chemical Identifiers
CAS | 10035-10-6, 7732-18-5 |
---|---|
Molecular Formula | BrH |
Molecular Weight (g/mol) | 80.91 |
MDL Number | MFCD00011323 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-N |
Synonym | hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide |
PubChem CID | 260 |
ChEBI | CHEBI:47266 |
IUPAC Name | hydrogen bromide |
SMILES | Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10025870
|
Thermo Scientific Acros
423785000 |
500 mL | Glass bottle |
76.80 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10761434
|
Thermo Scientific Acros
423780025 |
2.5 L | Glass bottle |
258.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
10035-10-6, 7732-18-5 | |
80.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
260 | |
hydrogen bromide |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CHEBI:47266 | |
Br |
Specifications
Colorless to Yellow | |
-11.0°C | |
1.4800g/mL | |
0.002% max. | |
BrH | |
Liquid | |
01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
hydrogen bromide | |
260 | |
80.9 | |
ACS Reagent |
10035-10-6, 7732-18-5 | |
126.0°C to 128.0°C | |
5ppm max. | |
1.48 | |
HBr | |
MFCD00011323 | |
15, 4816 | |
Solubility in water: soluble. | |
Br | |
80.91 | |
CHEBI:47266 | |
47.0 to 49.0% | |
Hydrobromic acid |
Safety and Handling
GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Danger
missing translation for 'einecsNumber' : 233-113-0
missing translation for 'rtecsNumber' : MW3850000
missing translation for 'tsca' : TSCA
- Hydrobromic acid, ca. 48 wt% solution in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only