missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic acid, Reagent Grade, 48%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Boiling Point | 126°C |
CAS Min % | ≥50.0000%, ≥30.0000% |
CAS Max % | <70.0000%, <50.0000% |
Density | 1.49 g/mL at 25 °C(lit.) |
Packaging | Glass Bottle |
Quantity | 500 mL |
Linear Formula | I2 |
UN Number | UN1788 |
Beilstein | 3587158 |
IUPAC Name | hydrogen bromide |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'