missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.1 N standard solution, Thermo Scientific Chemicals

29.95 € - 311.55 €

Specifications

Color Colorless
Name Note 0.1 N Standard Solution
CAS 7647-01-0, 7732-18-5
Boiling Point 100.0°C
Concentration 0.0998 to 0.1002N (20°C)
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10647282
View Documents
Thermo Scientific Acros
124200010
1 L Glass bottle
29.95 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10083943
View Documents
Thermo Scientific Acros
124200025
2.5 L Plastic bottle
49.95 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15294614
View Documents
Thermo Scientific Acros
124200100
10 L Plastic Drum
149.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
15204624
View Documents
Thermo Scientific Acros
124200250
25 L Plastic Can
311.55 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Specifications

Colorless
7647-01-0, 7732-18-5
0.0998 to 0.1002N (20°C)
1
HCl
MFCD00011324 MFCD00792839
15, 4818
Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene
Cl
313
36.46
0.1 N Standard Solution
100.0°C
1.0000g/mL
ClH
Liquid
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
36.46
CHEBI:17883
Videos
Safety and Handling

Safety and Handling

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated.
NFPA
Health: 2
Flammability: 0
Instability: 0

SIGNAL WORD: CAUTION!

Product Identifier
  • Hydrochloric acid, 0.1 N standard solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only