missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.5N Standardized Solution, Thermo Scientific Chemicals

31.00 € - 128.96 €

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333777
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
035642.K2
1 L
31.00 €
1 L
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
11323777
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
035642.K7
4 L
75.40 €
4 L
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
11313777
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
035642.M4
20 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description

Description

Hydrochloric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-01-0, 7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

Colorless liquid
7647-01-0, 7732-18-5
ClH
MFCD00011324 MFCD00792839
Miscible with water at room temperature. Also miscible with methanol,ethanol and ether.
Cl
36.46
CHEBI:17883
Hydrochloric acid: 4.13%; Water: 95.87%
Colorless
23 hPa (17mm Hg) at 20°C
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46
Hydrochloric acid
Videos
Safety and Handling

Safety and Handling

P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390

H290-H315-H318

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 231-595-7

missing translation for 'rtecsNumber' : MW4025000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only