missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1.0M, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Packaging:
1L
Description
Hydrochloric acid is a corrosive, strong mineral acid with many uses, including as a laboratory reagent in manual and automated titration. It is also used in chloride production, as a neutralizing agent for bases, as a catalyst and solvent in organic synthesis, and for HCl determination in the wine industry.
This solution is manufactured as per Official Methods of Analysis of AOAC International.
This solution is manufactured as per Official Methods of Analysis of AOAC International.
Specifications
Specifications
Color | Colorless |
CAS | 7647-01-0, 7732-18-5 |
Concentration | 1.0M |
Packaging | 1L |
Solvent or Matrix | Water |
Normality | 1.0N |
Quantity | 1 L |
Molecular Formula | ClH |
Physical Form | Liquid |
MDL Number | MFCD00011324 MFCD00792839 |
Show More |
Safety and Handling
Product Identifier
- HYDROCHLORIC ACID 1.0N (1.0M)
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : hydrochloric acid 37%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction