missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 1N standard solution, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 124210100


 View more versions of this product

Product Code. 15214624

  • 172.00 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hydrochloric acid
1N standard solution
100.0°C to 103.0°C
1.0200g/mL
1.02
ClH
Liquid
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
Colorless
7647-01-0, 7732-18-5
0.998 to 1.002N (20°C)
Plastic drum
10 L
HCl
MFCD00011324 MFCD00792839
15, 4818
Solubility in water: miscible. Other solubilities: soluble in alcohol and benzene
Cl
36.46
CHEBI:17883
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid, 1N standard solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only