missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 32%, Certified AR for Analysis, d=1.16, Fisher Chemical™
ClH, CAS Number-7647-01-0, spirits of salt, muriatic acid, anhydrous hydrochloric acid, hydrogen chloride hcl, acide chlorhydrique, hydrogen chloride, chloorwaterstof, hydrochloric acid, chlorohydric acid, chlorwasserstoff, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.45g/mol
33.75 € - 57.80 €
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | chlorane |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10264582
|
Fisher Chemical
H/1100/PB15 |
1 L |
33.75 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10458980
|
Fisher Chemical
H/1100/PB17 |
2.5 L |
57.80 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ClH, CAS Number-7647-01-0, spirits of salt, muriatic acid, anhydrous hydrochloric acid, hydrogen chloride hcl, acide chlorhydrique, hydrogen chloride, chloorwaterstof, hydrochloric acid, chlorohydric acid, chlorwasserstoff, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.45g/mol
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
1 | |
7647-01-0 | |
57°C | |
125mbar at 20°C | |
1.9 mPaS at 15°C | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.45g/mol |
Colorless | |
-35°C | |
HDPE plastic bottle | |
ClH | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |