missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid 36 - 38%, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Specifications
Melting Point | -30°C |
Boiling Point | 110°C (1013 hPa) |
Density | 1.18 g/cm³ (20°C) |
Residue after Ignition | max. 5 ppm |
Assay Percent Range | 37 to 38% |
Quantity | 1 L |
Safety and Handling
Product Identifier
- Hydrochloric acid 25-38%
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'