missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™

Product Code. 10496253
2.5 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

ClH, CAS Number-7647-01-0, anhydrous hydrochloric acid, acide chlorhydrique, chlorwasserstoff, hydrochloric acid, chloorwaterstof, chlorohydric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Packaging HDPE plastic bottle
Vapor Pressure 125mbar at 20°C
Quantity 2.5 L
Viscosity 1.8 mPaS at 15°C
Physical Form Liquid
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid 37%, for Trace Metal Analysis, d=1.18, Primar Plus™, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.