missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals

22.31 € - 80.70 €

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
SMILES [Cl-].[Cl-].[Pd++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11420570
View Documents
Thermo Scientific Alfa Aesar
043335.AP
500 mL
22.31 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11329508
View Documents
Thermo Scientific Alfa Aesar
043335.K2
1 L
33.06 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11319508
View Documents
Thermo Scientific Alfa Aesar
043335.K7
4 L
80.70 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-10-1
177.32
PIBWKRNGBLPSSY-UHFFFAOYSA-L
24290
[Cl-].[Cl-].[Pd++]
Cl2Pd
MFCD00003558
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
CHEBI:53434

Specifications

Colorless to Yellow
5% v/v aq. soln.
Liquid
UN1789
PIBWKRNGBLPSSY-UHFFFAOYSA-L
dichloropalladium
24290
36.46
7647-10-1
Cl2Pd
MFCD00003558
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
[Cl-].[Cl-].[Pd++]
177.32
CHEBI:53434
Hydrochloric acid
Videos
Safety and Handling

Safety and Handling

P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390

H290-H315-H318

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID

missing translation for 'einecsNumber' : 231-595-7

missing translation for 'rtecsNumber' : MW4025000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Hydrochloric acid, 5% v/v aqueous solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.