missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 99.999% (metals basis), 36.5% min, Thermo Scientific Chemicals

Product Code. 11324999
Click to view available options
Quantity:
250 mL
1000 mL
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl

Specifications

Appearance Colorless liquid
Color Colorless
Boiling Point 81.5°C to 110°C
Density 1.19
Odor Pungent
Assay Percent Range 99.999% (metals basis)
Quantity 2.5 L
Physical Form Liquid
Solubility Information Completely miscible with water.
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'