missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals Product Code.: 10167560

Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals

Product Code. 10167560
2.5 L, EcoSafPak™ Glass Bottle
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10167560

Brand: Thermo Scientific Acros 423790025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Density 1.1800g/mL
Heavy Metals (as Pb) 1ppm max.
Packaging EcoSafPak™ Glass Bottle
Residue after Ignition 0.0005% max.
Quantity 2.5 L
Linear Formula HCl
Physical Form Solution, Free From Suspended Matter or Sediment
UN Number 1789
Fieser 04,450; 05,533; 06,283
Merck Index 15, 4818
IUPAC Name chlorane
Formula Weight 36.45
Percent Purity 36.5 to 38.0%
Grade ACS Reagent
Chemical Name or Material Hydrochloric acid, ca. 37% Solution in Water
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.