missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, for analysis, ca. 32% solution in water, Thermo Scientific Chemicals

Product Code. 10083453
Click to view available options
Quantity:
2.5 L
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
CAS 7647-01-0, 7732-18-5
Melting Point -35.0°C
Boiling Point 57.0°C
Density 1.1600g/mL
Heavy Metals (as Pb) 1ppm max.
Packaging Glass bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.16
Quantity 2.5 L
Molecular Formula ClH
Viscosity 1.9 mPa.s (15°C)
Linear Formula HCl
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Merck Index 15, 4818
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Percent Purity ≥32%
Grade Analytical
Chemical Name or Material Hydrochloric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid, ca. 32% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'