missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid concentrate, For 1 L standard solution, 0.1 M HCl (0.1 N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Packaging | Ampoule |
Vapor Pressure | 190 mmHg (25°C) |
Quantity | 1 Pc. |
UN Number | UN1789 |
Formula Weight | 36.46g/mol |
Chemical Name or Material | Hydrochloric acid concentrate |
Safety and Handling
Product Identifier
- Hydrochloric acid concentrate
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'