missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate, For 1 L standard solution, 1.0 M HCl (1.0 N), Honeywell Fluka™

Product Code. 15675270
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.

for 1 L standard solution, 1.0 M HCl (1.0 N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Boiling Point 100°C
Concentration 1.0 M HCl (1.0 N)
CAS Max % 96.5000%, 3.5000%
Density 1.010 g/cm3
Packaging Ampoule
Vapor Pressure 190 mmHg (25°C)
Quantity 6 x 1 Ea.
Linear Formula HCl
Physical Form Concentrate
UN Number UN1789
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid concentrate, For 1 L standard solution, 1.0 M HCl (1.0 N)
Show More Show Less
compliance-icons
Product Identifier
  • Hydrochloric acid concentrate
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.