missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

ClH, CAS Number-7647-01-0, spirits of salt, hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride, chlorwasserstoff, hydrogen chloride hcl, muriatic acid, anhydrous hydrochloric acid, acide chlorhydrique, 6AMP, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol

36.50 € - 113.00 €

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10528040
View Documents
Fisher Chemical
J/4350C/05
1 Ampule
36.50 €
1 ampoule
Please to purchase this item. Need a web account? Register with us today!
10305760
View Documents
Fisher Chemical
J/4350C/90
6 Ampules This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

ClH, CAS Number-7647-01-0, spirits of salt, hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride, chlorwasserstoff, hydrogen chloride hcl, muriatic acid, anhydrous hydrochloric acid, acide chlorhydrique, 6AMP, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

Colorless
100°C
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46g/mol
7647-01-0
Ampoule
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Hydrochloric Acid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid, 0.1M (0.1N), concentrated solution=1M

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.