missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp
Click to view available options
Quantity:
1 L
2.5 L
Description
ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
Specifications
Specifications
pH | 2 |
Color | Colorless |
Boiling Point | 100°C |
Packaging | HDPE plastic bottle |
Vapor Pressure | 14mmHg @20°C |
Quantity | 1 L |
Physical Form | Liquid |
Formula Weight | 36.46g/mol |
Safety and Handling
Product Identifier
- Hydrochloric acid, 0.1M, (0.1N)
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction