missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Product Code. 10522533
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L

Hydrochloric Acid Solution 0.2M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

pH 2
Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Vapor Pressure 14mmHg @20°C
Quantity 1 L
Physical Form Liquid
IUPAC Name chlorane
Formula Weight 36.46g/mol
compliance-icons
Product Identifier
  • Hydrochloric acid, 0.2M, (0.2N)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'