missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, 0.2M, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
2 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Concentration | 0.2M |
Density | 1.005g/mL at 25°C |
Vapor Pressure | 190 mmHg (25°C) |
Quantity | 2 L |
UN Number | UN1789 |
IUPAC Name | chlorane |
Formula Weight | 36.46g/mol |
Chemical Name or Material | Hydrochloric acid solution |
Safety and Handling
Signal Word
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction