missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. 10590721
Click to view available options
Quantity:
1 Ampule
6 Ampules
This item is not returnable. View return policy
This item is not returnable. View return policy

ClH, CAS Number-7647-01-0, spirits of salt, hydrogen chloride, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof, acide chlorhydrique, chlorohydric acid, muriatic acid, chlorwasserstoff, hydrochloric acid, 6AMP, CHEBI:17883, Colorless to Yellow, VEXZGXHMUGYJMC-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

pH lt; 1
Color Colorless to Yellow
Packaging Ampoule
Quantity 1 Ampule
Physical Form Liquid
IUPAC Name chlorane
compliance-icons compliance-icons
Product Identifier
  • 0.5M (0.5N) Hydrochloric acid (5M Concentrate for volumetric analysis)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'