missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™ Product Code.: 10386040

Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™

Product Code. 10386040
1 Ampule, Ampoule
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ampule
Packaging:
Ampoule
This item is not returnable. View return policy

Product Code. 10386040

Brand: Fisher Chemical J/4320C/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For volumetric analysis
TRUSTED_SUSTAINABILITY

Specifications

pH 1
Color Colorless
CAS 7647-01-0
Melting Point -35°C
Boiling Point 57°C
Packaging Ampoule
Vapor Pressure 125mbar at 20°C
Quantity 1 Ampule
Molecular Formula ClH
Viscosity 1.9 mPaS at 15°C
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46 g/mol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528095 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberJ/4320C/05,J/4320/99,J/4320C/90
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid, 1M, (1N) concentrated solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.