missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™
Click to view available options
Quantity:
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
Description
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis
Specifications
Specifications
Color | Colorless |
Boiling Point | 100°C |
Packaging | HDPE plastic bottle |
Quantity | 2.5 L |
Physical Form | Liquid |
IUPAC Name | chlorane |
Formula Weight | 36.46g/mol |
Safety and Handling
Product Identifier
- Hydrochloric acid, 1M, (1N)
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction