missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 1 M HCl (1N), Honeywell Fluka™

Product Code. 15634940
Click to view available options
Quantity:
1 L
5 L
10 L
20 L
30 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
Plastic Pail
VOLPAC™ CONTAINER
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 1 M HCl (1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Vapor Pressure 190 mmHg (25°C)
Quantity 10 L
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46
Grade Reagent Ph. Eur.
Chemical Name or Material Hydrochloric acid solution, volumetric, 1 M HCl (1N)
compliance-icons
Product Identifier
  • Hydrochloric acid solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'