missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 1 M HCl (1N), Honeywell Fluka™

Product Code. 15664940
Click to view available options
Quantity:
1 L
5 L
10 L
20 L
30 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
Plastic Pail
VOLPAC™ CONTAINER
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, Reag. Ph. Eur., 1 M HCl (1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Concentration 1M
Vapor Pressure 190 mmHg (25°C)
Quantity 20 L
UN Number UN1789
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid solution
compliance-icons
Product Identifier
  • Hydrochloric acid solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.