missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 5M (5N), Standard Solution ready to use for volumetric analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
Description
Hydrochloric Acid Solution 5M (5N), Standard Solution ready to use for volumetric analysis
Specifications
Specifications
pH | lt; 1 |
Color | Colorless to Yellow |
Packaging | HDPE plastic bottle |
Quantity | 2.5 L |
Physical Form | Liquid |
IUPAC Name | hydrogen chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'